2'-Trifluoromethoxyacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-[2-(Trifluoromethoxy)phenyl]-1-ethanone, 1-(2-(Trifluoromethoxy)phenyl)ethanone

006484-5G 006484-25G
101790-602EA 60 USD
101790-602 101790-604
2'-Trifluoromethoxyacetophenone ≥98%
2'-Trifluoromethoxyacetophenone

Hazard code(s): Irritant

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
MDL Number: MFCD00236320
CAS Number: 220227-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR