2,4'-Difluorobenzophenone ≥97%

Supplier: Matrix Scientific

Synonyms: (2-Fluorophenyl)(4-fluorophenyl)methanone

006466-5G 006466-25G
101790-588EA 21.54 USD
101790-588 102550-872
2,4'-Difluorobenzophenone ≥97%
2,4'-Difluorobenzophenone

Hazard code(s): Irritant

Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 22 - 24 °C
Density: 1.24 g/cm³ (20 °C)
MDL Number: MFCD00000319
CAS Number: 342-25-6
EINECS: 206-441-7

Order Now

Specification Test Results

Boiling point 176-178°C/1
Melting point 22-24°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR