4,4'-Difluorobenzhydrol ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 4,4'-Difluorophenyl phenyl carbinol, 4,4'-Difluorobenzhydryl alcohol, 4,4'-Difluoro-α-phenylbenzyl alcohol, Bis(4-fluorophenyl)carbinol, 4,4'-Difluorodiphenylmethanol, Bis(4-fluorophenyl)methanol

006462-25G 006462-100G 006462-500G
101790-580EA 69.23 USD
101790-580 101790-582 101790-584
4,4'-Difluorobenzhydrol ≥98% for synthesis
4,4'-Difluorobenzhydrol

Hazard code(s): Irritant

Formula: C₁₃H₁₀F₂O
MW: 220.22 g/mol
Boiling Pt: 305
Melting Pt: 47
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00000357
CAS Number: 365-24-2
EINECS: 206-671-8

Order Now

Specification Test Results

Boiling point 143°C/3mm
Melting point 47-48°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR