2-Bromofluorene ≥97% for synthesis

Supplier: Matrix Scientific
Warning

006441-25G 006441-100G
101790-568EA 75.38 USD
101790-568 102550-868
2-Bromofluorene ≥97% for synthesis
2-Bromofluorene

Hazard code(s): Irritant

Formula: C₁₃H₉Br
Boiling Pt: 185 °C (135mm)
Melting Pt: 110…114 °C
MDL Number: MFCD00001115
CAS Number: 1133-80-8
EINECS: 214-480-6

Order Now

Specification Test Results

Boiling point 185°C/135mm
Melting point 112-114°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR