8-Methylquinoline ≥97%

Supplier: Matrix Scientific
Warning

006429-5G 006429-25G 006429-100G
101790-556EA 18.46 USD
101790-556 101790-558 101790-560
8-Methylquinoline ≥97%
8-Methylquinoline

Hazard code(s): Irritant

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 135…136 °C (25mm)
Melting Pt: –80 °C
Density: 1.0852 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006810
CAS Number: 611-32-5
EINECS: 210-264-0

Order Now

Specification Test Results

Boiling point 135-136°C/2
Melting point -80°C
Density 1.052
ND20 1.6145

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR