7-Chloroquinaldine ≥99%

Supplier: Matrix Scientific

Synonyms: 7-Chloro-2-methylquinoline

006424-5G 006424-25G 006424-100G
101790-540EA 21.54 USD
101790-540 101790-542 102663-562
7-Chloroquinaldine ≥99%
7-Chloroquinaldine

Hazard code(s): Irritant

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Melting Pt: 74…78 °C
MDL Number: MFCD00075436
CAS Number: 4965-33-7
EINECS: 440-600-3

Order Now

Specification Test Results

Melting point 74-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR