Ethyl-3-(trifluoromethyl)benzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-(trifluoromethyl)benzoate

006386-5G 006386-25G
101790-482EA 41.54 USD
101790-482 101790-484
Ethyl-3-(trifluoromethyl)benzoate ≥97%
Ethyl-3-(trifluoromethyl)benzoate

Hazard code(s): Irritant

Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Boiling Pt: 100…101 °C (10mm)
MDL Number: MFCD00013560
CAS Number: 76783-59-0

Order Now

Specification Test Results

Boiling point 100-101°C/1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR