3,4'-Bis(trifluoromethyl)benzophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(Trifluormethyl)-3-((4-(trifluormethyl)phenyl)methanoyl)benzene

006373-5G 006373-25G
101790-440EA 70.77 USD
101790-440 101790-442
3,4'-Bis(trifluoromethyl)benzophenone ≥98%
3,4'-Bis(trifluoromethyl)benzophenone

Hazard code(s): Irritant

Formula: C₁₅H₈F₆O
MW: 318.22 g/mol
Melting Pt: 96…97 °C
MDL Number: MFCD00013561
CAS Number: 21084-22-0

Order Now

Specification Test Results

Melting point 96-97°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR