2-(Methylthio)aniline ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(Methylmercapto)aniline
006301-25G
006301-100G
101790-354EA
100
USD
101790-354
101790-356
2-(Methylthio)aniline ≥97%
2-(Methylthio)aniline
Hazard code(s): Toxic, Stench
Formula:
C₇H₉NS MW: 139.22 g/mol Boiling Pt: 234 °C Density: 1.1333 g/cm³ (17 °C) |
MDL Number:
MFCD00007708 CAS Number: 2987-53-3 EINECS: 221-055-9 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 234°C |
Density | 1.111 |
ND20 | 1.624 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...