Pentifylline ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 1-Hexyl-3,7-dimethyl-1H-purine-2,6(3H,7H)-dione, 1-Hexyl-3,7-dimethylxanthine, 3,7-Dimethyl-1-hexylxanthine, 1-Hexyltheobromine, 1-n-Hexyltheobromine, 2,6-Dihydroxy-3,7-dimethyl-1-hexylpurine

006295-1G 006295-5G
101790-340EA 75.38 USD
101790-340 101790-342
Pentifylline ≥98% for synthesis
Pentifylline

Hazard code(s): Irritant-Harmful

Formula: C₁₃H₂₀N₄O₂
MW: 264.33 g/mol
Melting Pt: 82…83 °C
MDL Number: MFCD00041424
CAS Number: 1028-33-7
EINECS: 213-842-0

Order Now

Specification Test Results

Melting point 82-83°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR