2,2,3,3,3-Pentafluoro-1-propanol ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 1H,1H-Pentafluoro-1-propanol, PFPOH (pentafluoropropanol), 1H,1H-Pentafluoropropanol-1, 2,2,3,3,3-Pentafluoropropanol

006277-25G 006277-100G
101790-318EA 40 USD
101790-318 101790-320
2,2,3,3,3-Pentafluoro-1-propanol ≥97% for synthesis
2,2,3,3,3-Pentafluoro-1-propanol

Hazard code(s): Irritant

Formula: C₃H₃F₅O
Boiling Pt: 78 - 80 °C (1013 hPa)
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00004673
CAS Number: 422-05-9
EINECS: 207-012-7

Order Now

Specification Test Results

Boiling point 81°C
Density 1.509
ND20 1.288


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR