1,1,1,2,2,3,3-Heptafluoro-4-iodobutane ≥97%

Supplier: Matrix Scientific

Synonyms: 1H,1H-Heptafluorobutyl iodide, 2,2,3,3,4,4,4-Heptafluoro-1-iodobutane

006265-1G 006265-5G
101790-300EA 87.69 USD
101790-300 101790-302
1,1,1,2,2,3,3-Heptafluoro-4-iodobutane ≥97%
1,1,1,2,2,3,3-Heptafluoro-4-iodobutane

Hazard code(s): Irritant

Formula: C₄H₂F₇I
MW: 309.95 g/mol
Boiling Pt: 91…92 °C
Density: 2.054 g/cm³
MDL Number: MFCD00013719
CAS Number: 374-98-1
EINECS: 206-779-5

Order Now

Specification Test Results

Boiling point 91-92°C
Density 2.054
ND20 1.367

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR