1H,1H-Heptafluorobutyl acetate ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2,3,3,4,4,4-heptafluorobutyl acetate

006254-5G 006254-25G
101790-274EA 92.31 USD
101790-274 101790-276
1H,1H-Heptafluorobutyl acetate ≥97%
1H,1H-Heptafluorobutyl acetate

Hazard code(s): Irritant

Formula: C₆H₅F₇O₂
MW: 242.09 g/mol
Boiling Pt: 105 °C (735mm)
Density: 1.435 g/cm³
MDL Number: MFCD01070838
CAS Number: 356-06-9

Order Now

Specification Test Results

Boiling point 105°C/735mm
Density 1.435
ND20 1.311

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR