1H,1H-Pentadecafluoro-1-octanol ≥98%

Supplier: Matrix Scientific

Synonyms: 1H,1H-Perfluoro-1-octanol, 1H,1h-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluor-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluoro-1-octanol

006250-25G 006250-100G
101790-262EA 45.95 USD
101790-262 101790-264 101790-266
1H,1H-Pentadecafluoro-1-octanol ≥98%
1H,1H-Pentadecafluoro-1-octanol
Formula: C₈H₃F₁₅O
MW: 400.09 g/mol
MDL Number: MFCD00004675
CAS Number: 307-30-2
EINECS: 206-197-1

Order Now

Specification Test Results

Boiling point 163-165°C
Melting point 36-37°C
Density 1.388
ND20 1.3084

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR