4-(Methylsulfonyl)benzyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: α-Chloro-4-(methylsulphonyl)toluene, 1-(Chloromethyl)-4-(methylsulphonyl)benzene

006227-1G 006227-5G
101790-254EA 60 USD
101790-254 101790-256
4-(Methylsulfonyl)benzyl chloride ≥98%
4-(Methylsulphonyl)benzyl chloride

Hazard code(s): Irritant, Lachrymator

Formula: C₈H₉ClO₂S
MW: 204.68 g/mol
Melting Pt: 78…80 °C
MDL Number: MFCD00079774
CAS Number: 40517-43-9

Order Now

Specification Test Results

Melting point 78-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR