1-Pentanesulphonic acid sodium salt ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate

006219-5G 006219-25G
101790-246EA 556.92 USD
101790-246 101790-248
1-Pentanesulphonic acid sodium salt ≥98% for synthesis
1-Pentanesulphonic acid sodium salt

Hazard code(s): Irritant

Formula: C₅H₁₁O₃SNa
Melting Pt: ? 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
EINECS: 245-208-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Melting point >300°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR