4-(Methylthio)phenylboronic acid ≥97%

Supplier: Matrix Scientific

006203-5G 006203-25G
101790-234EA 53.85 USD
101790-234 101790-236
4-(Methylthio)phenylboronic acid ≥97%
4-(Methylthio)phenylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₇H₉BO₂S
Melting Pt: 210…212 °C
MDL Number: MFCD00093410
CAS Number: 98546-51-1

Order Now

Specification Test Results

Melting point 210-212°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR