3,5-Dihydroxybenzhydrazide

Supplier: Matrix Scientific

Synonyms: 3,5-Dihydroxybenzoic hydrazide

006161-5G 006161-25G
101790-176EA 87.69 USD
101790-176 101790-178
3,5-Dihydroxybenzhydrazide
3,5-Dihydroxybenzhydrazide

Hazard code(s): Irritant

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Melting Pt: 265 °C (dec.)
MDL Number: MFCD00017066
CAS Number: 7732-32-3

Order Now

Specification Test Results

Melting point 265°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR