3,4-Dihydroxybenzhydrazide ≥98%

Supplier: Matrix Scientific

006160-5G 006160-25G
101790-172EA 100 USD
101790-172 101790-174
3,4-Dihydroxybenzhydrazide ≥98%
3,4-Dihydroxybenzhydrazide

Hazard code(s): Irritant

Formula: C₇H₈N₂O₃
Melting Pt: 268 °C
MDL Number: MFCD00017064
CAS Number: 39635-11-5
EINECS: 254-550-3

Order Now

Specification Test Results

Melting point 268°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR