3-(3-Fluorophenyl)propionic acid ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-(3-Fluorophenyl)propanoic acid

006112-1G 006112-5G 006112-25G
101790-116EA 44.62 USD
101790-116 101790-118 101790-120
3-(3-Fluorophenyl)propionic acid ≥98% for synthesis
3-(3-Fluorophenyl)propionic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₉H₉FO₂
MW: 168.17
Boiling Pt: 125 °C (10mm)
Melting Pt: 40…42 °C
MDL Number: MFCD01310821
CAS Number: 458-45-7

Order Now

Specification Test Results

Boiling point 125°C/10mm
Melting point 40-42°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR