3-(2-Fluorophenyl)propionic acid ≥98%

Supplier: Matrix Scientific

006111-1G 006111-5G 006111-25G
101790-110EA 15.38 USD
101790-110 101790-112 101790-114
3-(2-Fluorophenyl)propionic acid ≥98%
3-(2-Fluorophenyl)propionic acid

Hazard code(s): Irritant

Formula: C₉H₉FO₂
Melting Pt: 76…78 °C
MDL Number: MFCD01310820
CAS Number: 1643-26-1

Order Now

Specification Test Results

Melting point 76-78°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR