3-(3-Chlorophenyl)propionic acid ≥98%
Supplier: Matrix Scientific
006109-1G
006109-5G
006109-25G
101790-100EA
16.92
USD
101790-100
101790-102
101790-104
3-(3-Chlorophenyl)propionic acid ≥98%
3-(3-Chlorophenyl)propionic acid
Hazard code(s): Irritant
Formula:
C₉H₉ClO₂ Melting Pt: 72…74 °C |
MDL Number:
MFCD01310801 CAS Number: 21640-48-2 |
Specification Test Results
Melting point | 72-74°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...