3-(2-Chlorophenyl)propionic acid ≥98%

Supplier: Matrix Scientific

006108-1G 006108-5G
101790-096EA 15.38 USD
101790-096 101790-098
3-(2-Chlorophenyl)propionic acid ≥98%
3-(2-Chlorophenyl)propionic acid

Hazard code(s): Irritant

Formula: C₉H₉ClO₂
Melting Pt: 94…96 °C
MDL Number: MFCD00016547
CAS Number: 1643-28-3

Order Now

Specification Test Results

Melting point 94-96°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR