1-(4-Fluorophenyl)imidazole ≥98%

Supplier: Matrix Scientific

006101-1G 006101-5G
101790-082EA 106.15 USD
101790-082 101790-084
1-(4-Fluorophenyl)imidazole ≥98%
1-(4-Fluorophenyl)imidazole

Hazard code(s): Irritant

Formula: C₉H₇FN₂
Boiling Pt: 80 °C (0.3mm)
MDL Number: MFCD00060490
CAS Number: 21441-24-7

Order Now

Specification Test Results

Boiling point 80°C/0.3mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR