1-(2-Chlorophenyl)imidazole ≥99%

Supplier: Matrix Scientific

006098-1G 006098-5G
101790-070EA 150.77 USD
101790-070 101790-072
1-(2-Chlorophenyl)imidazole ≥99%
1-(2-Chlorophenyl)imidazole

Hazard code(s): Irritant

Formula: C₉H₇ClN₂
Boiling Pt: 108 °C (1mm)
Density: 1.25 g/cm³
MDL Number: MFCD00041205
CAS Number: 51581-50-1

Order Now

Specification Test Results

Boiling point 108°C/1mm
Density 1.25
ND20 1.59


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR