1-(4-Chlorophenyl)imidazole ≥98%
Supplier: Matrix Scientific
006092-1G
006092-5G
101790-046EA
120
USD
101790-046
101790-048
1-(4-Chlorophenyl)imidazole ≥98%
1-(4-Chlorophenyl)imidazole
Hazard code(s): Irritant
Formula:
C₉H₇ClN₂ Melting Pt: 85…87 °C |
MDL Number:
MFCD00041208 CAS Number: 51581-54-5 |
Specification Test Results
Melting point | 85-87°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...