1-(4-Fluorophenyl)-2-thiourea ≥99%
Supplier: Matrix Scientific
|
Danger
|
Synonyms:
N-(4-Fluorophenyl)thiourea, 4-Fluorophenylthiourea
006078-5G
006078-25G
101790-024EA
21.54
USD
101790-024
101790-026
1-(4-Fluorophenyl)-2-thiourea ≥99%
1-(4-Fluorophenyl)-2-thiourea
Hazard code(s): Irritant
Formula:
(C₆H₄F)NHC(S)NH₂ MW: 170.21 g/mol Boiling Pt: 276 °C (1013 hPa) Melting Pt: 169 - 170 °C |
MDL Number:
MFCD00041180 CAS Number: 459-05-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 163-164°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...