1-Bromo-4-(difluoromethoxy)benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Difluoromethoxy)bromobenzene

006023-5G 006023-25G 006023-100G
101789-938EA 24.62 USD
101789-938 101789-940 101789-942
1-Bromo-4-(difluoromethoxy)benzene ≥97%
1-Bromo-4-(difluoromethoxy)benzene

Hazard code(s): Irritant

Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
Boiling Pt: 96…98 °C (15mm)
MDL Number: MFCD00221465
CAS Number: 5905-69-1

Order Now

Specification Test Results

Boiling point 96-98°C/15m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR