1-Bromo-2-(difluoromethoxy)benzene ≥97%

Supplier: Matrix Scientific

006021-5G 006021-25G
101789-930EA 26.15 USD
101789-930 101789-932
1-Bromo-2-(difluoromethoxy)benzene ≥97%
1-Bromo-2-(difluoromethoxy)benzene

Hazard code(s): Irritant

Formula: C₇H₅BrF₂O
Boiling Pt: 85…87 °C (15mm)
MDL Number: MFCD00221468
CAS Number: 175278-33-8

Order Now

Specification Test Results

Boiling point 85-87°C/15m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR