1-(Difluoromethoxy)-2-nitrobenzene ≥97% for synthesis

Supplier: Matrix Scientific

006020-1G 006020-5G 006020-25G
101789-924EA 41.54 USD
101789-924 101789-926 101789-928
1-(Difluoromethoxy)-2-nitrobenzene ≥97% for synthesis
1-(Difluoromethoxy)-2-nitrobenzene

Hazard code(s): Irritant

Formula: C₇H₅F₂NO₃
Boiling Pt: 73…75 °C (0.3mm)
MDL Number: MFCD00221470
CAS Number: 22225-77-0

Order Now

Specification Test Results

Boiling point 73-75°C/0.3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR