2-(Difluoromethoxy)benzyl alcohol
Supplier: Matrix Scientific
006002-1G
006002-5G
101789-900EA
60
USD
101789-900
101789-902
2-(Difluoromethoxy)benzyl alcohol
2-(Difluoromethoxy)benzyl alcohol
Formula:
C₈H₈F₂O₂ MW: 174.15 g/mol |
MDL Number:
MFCD00236226 CAS Number: 72768-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...