1-(Difluoromethoxy)-4-nitrobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Difluoromethoxy)nitrobenzene

005997-1G 005997-5G 005997-25G
101789-890EA 15.38 USD
101789-890 101789-892 101789-894
1-(Difluoromethoxy)-4-nitrobenzene ≥98%
1-(Difluoromethoxy)-4-nitrobenzene

Hazard code(s): Irritant

Formula: C₇H₅F₂NO₃
MW: 189.12 g/mol
Melting Pt: 33…35 °C
MDL Number: MFCD00085009
CAS Number: 1544-86-1

Order Now

Specification Test Results

Melting point 33-35°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR