3-(4-Fluorobenzoyl)propionic acid ≥97%
Supplier: Matrix Scientific
005951-5G
005951-25G
101789-836EA
27.69
USD
101789-836
101789-838
3-(4-Fluorobenzoyl)propionic acid ≥97%
3-(4-Fluorobenzoyl)propionic acid
Hazard code(s): Irritant
Formula:
C₁₀H₉FO₃ MW: 196.18 g/mol Melting Pt: 100…102 °C |
MDL Number:
MFCD00002793 CAS Number: 366-77-8 EINECS: 206-679-1 |
Specification Test Results
Melting point | 100-102°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...