α,α,α-Trifluoro-3,5-dinitrotoluene ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3,5-Dinitrobenzotrifluoride

005948-25G 005948-100G
101789-828EA 26.15 USD
101789-828 101789-830
α,α,α-Trifluoro-3,5-dinitrotoluene ≥98%
α,α,α-Trifluoro-3,5-dinitrotoluene

Hazard code(s): Irritant

Formula: C₇H₃F₃N₂O₄
MW: 236.11 g/mol
Boiling Pt: 127 - 129 °C (9 -rr)
Melting Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00007233
CAS Number: 401-99-0
EINECS: 206-935-2

Order Now

Specification Test Results

Boiling point 125-128°C/9
Melting point 49-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR