Tetrafluorohydroquinone ≥98%
Supplier: Matrix Scientific
Synonyms:
2,3,5,6-Tetrafluorohydroquinone, Tetrafluorobenzene-1,4-diol
005940-5G
005940-25G
101789-800EA
92.31
USD
101789-800
101789-802
Tetrafluorohydroquinone ≥98%
Tetrafluorohydroquinone
Hazard code(s): Irritant
Formula:
C₆H₂F₄O₂ MW: 182.07 g/mol Melting Pt: 168…170 °C |
MDL Number:
MFCD00002340 CAS Number: 771-63-1 EINECS: 212-237-9 |
Specification Test Results
Melting point | 168-170°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...