Ethyl nitroacetate ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: Nitroethansäureethylester, Nitroacetic ethyl ester

005934-5G 005934-25G 005934-100G
101789-784EA 15.38 USD
101789-784 101789-786 101789-788
Ethyl nitroacetate ≥95% for synthesis
Ethyl nitroacetate

Hazard code(s): Irritant, Refrigerate

Formula: C₄H₇NO₄
Boiling Pt: 93 - 95 °C (13.3 hPa)
Density: 1.226 g/cm³ (0 °C)
MDL Number: MFCD00007403
CAS Number: 626-35-7
EINECS: 210-944-7

Order Now

Specification Test Results

Boiling point 93-95°C/10m
Density 1.199


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR