3,5-Bis(trifluoromethyl)benzaldehyde ≥95%

Supplier: Matrix Scientific

005873-25G 005873-100G
101789-680EA 46.15 USD
101789-680 101789-682
3,5-Bis(trifluoromethyl)benzaldehyde ≥95%
3,5-Bis(trifluoromethyl)benzaldehyde

Hazard code(s): Irritant

Formula: C₉H₄F₆O
MW: 242.12 g/mol
Boiling Pt: 37 °C (1.3mm)
Melting Pt: –2 °C
Density: 1.46 g/cm³ (20 °C)
MDL Number: MFCD00010206
CAS Number: 401-95-6

Order Now

Specification Test Results

Boiling point 37°C/1.3mm
Density 1.469
ND20 1.422

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR