(2,4-Bis(trifluoromethyl)phenyl)acetic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2,4-Bis(trifluoromethyl)phenylacetic acid

005840-1G 005840-5G
101789-624EA 63.08 USD
101789-624 101789-626
(2,4-Bis(trifluoromethyl)phenyl)acetic acid ≥97%
(2,4-Bis(trifluoromethyl)phenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃(CF₃)₂)OCH₂COOH
MW: 272.15 g/mol
MDL Number: MFCD00042492
CAS Number: 177952-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR