2,4-Bis(trifluoromethyl)benzyl alcohol ≥98% for synthesis

Supplier: Matrix Scientific

005827-1G 005827-5G
101789-600EA 44.62 USD
101789-600 101789-602
2,4-Bis(trifluoromethyl)benzyl alcohol ≥98% for synthesis
2,4-Bis(trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant

Formula: C₉H₆F₆O
MDL Number: MFCD00042491
CAS Number: 143158-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR