2,4-Bis(trifluoromethyl)benzaldehyde ≥96%

Supplier: Matrix Scientific

005817-5G 005817-25G
101789-584EA 172.31 USD
101789-584 101789-586
2,4-Bis(trifluoromethyl)benzaldehyde ≥96%
2,4-Bis(trifluoromethyl)benzaldehyde

Hazard code(s): Irritant

Formula: C₉H₄F₆O
MW: 242.12 g/mol
Boiling Pt: 172 °C
MDL Number: MFCD00083535
CAS Number: 59664-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR