(3,5-Bis(trifluoromethyl)phenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

005810-5G 005810-25G 005810-100G
101789-568EA 15.38 USD
101789-568 101789-570 101789-572
(3,5-Bis(trifluoromethyl)phenyl)acetic acid ≥98%
(3,5-Bis(trifluoromethyl)phenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃(CF₃)₂)CH₂COOH
MW: 272.15 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 121…123 °C
MDL Number: MFCD00009908
CAS Number: 85068-33-3
EINECS: 285-295-6

Order Now

Specification Test Results

Melting point 121-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR