3,5-Bis(trifluoromethyl)benzyl alcohol ≥98%

Supplier: Matrix Scientific

Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanol

005808-5G 005808-25G 005808-100G
101789-562EA 16.92 USD
101789-562 101789-564 101789-566
3,5-Bis(trifluoromethyl)benzyl alcohol ≥98%
3,5-Bis(trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant

Formula: C₉H₆F₆O
MW: 244.14 g/mol
Boiling Pt: 100 °C (14 mmHg)
Melting Pt: 54…56 °C
MDL Number: MFCD00009907
CAS Number: 32707-89-4
EINECS: 251-168-9

Order Now

Specification Test Results

Melting point 54-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR