3,5-Bis(trifluoromethyl)benzoic acid ≥98%

Supplier: Matrix Scientific

005807-100G 005807-500G
101789-558EA 49.23 USD
101789-558 101789-560
3,5-Bis(trifluoromethyl)benzoic acid ≥98%
3,5-Bis(trifluoromethyl)benzoic acid

Hazard code(s): Irritant

Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 142…143 °C (subl.)
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00000388
CAS Number: 725-89-3
EINECS: 211-970-1

Order Now

Specification Test Results

Melting point 142-143°C(s

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR