Methyl pentadecafluorooctanoate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl pentadecafluorooctanoate, Methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate, Methyl pentadecafluorocaprylate, Pentadecafluorooctanoic acid methyl ester, Pentadecafluorocaprylic acid methyl ester, Methyl perfluorooctanoate

005790-5G 005790-25G
101789-538EA 35.38 USD
101789-538 101789-540
Methyl pentadecafluorooctanoate ≥98%
Methyl pentadecafluorooctanoate

Hazard code(s): Irritant

Formula: C₉H₃F₁₅O₂
MW: 428.1 g/mol
Boiling Pt: 159…160 °C
Density: 1.786 g/cm³
MDL Number: MFCD00039244
CAS Number: 376-27-2
EINECS: 206-808-1

Order Now

Specification Test Results

Boiling point 159-160°C
Density 1.786
ND20 1.305

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR