2'-Fluoropropiophenone ≥99%

Supplier: Matrix Scientific

Synonyms: o-Fluoropropiophenone

005743-25G 005743-100G
101789-472EA 36.92 USD
101789-472 101789-474
2'-Fluoropropiophenone ≥99%
2'-Fluoropropiophenone

Hazard code(s): Irritant

Formula: C₉H₉FO
MW: 152.17 g/mol
Boiling Pt: 89…91 °C (12mm)
Density: 1.102 g/cm³
MDL Number: MFCD00009893
CAS Number: 446-22-0

Order Now

Specification Test Results

Boiling point 89-91°C/12m
Density 1.102
ND20 1.504

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR