3'-Fluoropropiophenone ≥97%

Supplier: Matrix Scientific

Synonyms: m-Fluoropropiophenone, 1-(3-Fluorophenyl)propan-1-one

005740-1G 005740-5G 005740-25G
101789-466EA 20 USD
101789-466 101789-468 101789-470
3'-Fluoropropiophenone ≥97%
3'-Fluoropropiophenone

Hazard code(s): Irritant

Formula: C₉H₉FO
MW: 152.17 g/mol
MDL Number: MFCD00061245
CAS Number: 455-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR