4-Methoxyphenylboronic acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
005719-25G
005719-100G
101789-432EA
69.23
USD
101789-432
101789-434
4-Methoxyphenylboronic acid ≥97%
4-Methoxyphenylboronic acid
Hazard code(s): Irritant, Store in Freezer
Formula:
C₇H₉BO₃ Melting Pt: 204 - 207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039139 CAS Number: 5720-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...