3-Methoxyphenylboronic acid ≥97%

Supplier: Matrix Scientific
Warning

005718-25G 005718-100G
101789-428EA 49.23 USD
101789-428 101789-430
3-Methoxyphenylboronic acid ≥97%
3-Methoxyphenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₇H₉BO₃
Melting Pt: 160…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00161359
CAS Number: 10365-98-7

Order Now

Specification Test Results

Melting point 160-162°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR