(3-Phenoxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

005709-1G 005709-5G
101789-396EA 41.54 USD
101789-396 101789-398
(3-Phenoxyphenyl)acetic acid ≥98%
(3-Phenoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₅OC₆H₅)CH₂COOH
Melting Pt: 87…89 °C
MDL Number: MFCD00016826
CAS Number: 32852-81-6

Order Now

Specification Test Results

Melting point 87-89°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR