(2-Phenoxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific

005708-1G 005708-5G
101789-392EA 120 USD
101789-392 101789-394
(2-Phenoxyphenyl)acetic acid ≥98%
(2-Phenoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₅OC₆H₄)CH₂COOH
MW: 242.27 g/mol
Melting Pt: 89…91 °C
MDL Number: MFCD00079778
CAS Number: 25563-02-4

Order Now

Specification Test Results

Melting point 89-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR